RIE156320597 Rieke Metals, Inc.

3-Acetoxy-4'-t-butylbenzophenone

  • MDL number MFCD07698895
  • Cas Number : 890099-86-2
  • Molecular Weight : 296.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5176-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5176-34
Availability details for
Quantity
  • Availability
  • Add to Cart