RIE156320606 Rieke Metals, Inc.

3-Acetoxy-4'-butoxybenzophenone

  • MDL number MFCD07698904
  • Cas Number : 890100-04-6
  • Molecular Weight : 312.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5176-36C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5176-36C
Availability details for
Quantity
  • Availability
  • Add to Cart