RIE156320618 Rieke Metals, Inc.

3-Acetoxy-2',5'-dichlorobenzophenone

  • MDL number MFCD07698916
  • Cas Number : 890100-28-4
  • Molecular Weight : 309.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5176-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5176-45
Availability details for
Quantity
  • Availability
  • Add to Cart