RIE156320627 Rieke Metals, Inc.

3-Acetoxy-3',5'-dimethoxybenzophenone

  • MDL number MFCD07698925
  • Cas Number : 890099-02-2
  • Molecular Weight : 300.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5176-54

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5176-54
Availability details for
Quantity
  • Availability
  • Add to Cart