RIE156320633 Rieke Metals, Inc.

3-Acetoxy-3',5'-dimethylbenzophenone

  • MDL number MFCD07698931
  • Cas Number : 890099-20-4
  • Molecular Weight : 268.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5176-60

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5176-60
Availability details for
Quantity
  • Availability
  • Add to Cart