RIE156320636 Rieke Metals, Inc.

4-Acetoxy-2'-methoxybenzophenone

  • MDL number MFCD07698935
  • Cas Number : 124208-72-6
  • Molecular Weight : 270.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5177-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-02
Availability details for
Quantity
  • Availability
  • Add to Cart