RIE156320642 Rieke Metals, Inc.

4-Acetoxy-2'-trifluoromethylbenzophenone

  • MDL number MFCD07698941
  • Cas Number : 890099-32-8
  • Molecular Weight : 308.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5177-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-08
Availability details for
Quantity
  • Availability
  • Add to Cart