RIE156320652 Rieke Metals, Inc.

4-Acetoxy-4'-bromobenzophenone

  • MDL number MFCD07698952
  • Cas Number : 4306-46-1
  • Molecular Weight : 319.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(4-bromobenzoyl)phenyl acetate
Inchi Code 1S/C15H11BrO3/c1-10(17)19-14-8-4-12(5-9-14)15(18)11-2-6-13(16)7-3-11/h2-9H,1H3
Inchi Key IFRAFNJMZOSYKW-UHFFFAOYSA-N

Description

Purity 90%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5177-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-19
Availability details for
Quantity
  • Availability
  • Add to Cart