RIE156320655 Rieke Metals, Inc.

4-Acetoxy-4'-iodobenzophenone

  • MDL number MFCD07698955
  • Cas Number : 890099-52-2
  • Molecular Weight : 366.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(4-iodobenzoyl)phenyl acetate
Inchi Code 1S/C15H11IO3/c1-10(17)19-14-8-4-12(5-9-14)15(18)11-2-6-13(16)7-3-11/h2-9H,1H3
Inchi Key ONTOVLJRCHFHEJ-UHFFFAOYSA-N

Description

Purity 90%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5177-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-22
Availability details for
Quantity
  • Availability
  • Add to Cart