RIE156320657 Rieke Metals, Inc.

4-Acetoxy-4'-cyanobenzophenone

  • MDL number MFCD07698957
  • Cas Number : 890099-56-6
  • Molecular Weight : 265.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5177-25

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-25
Availability details for
Quantity
  • Availability
  • Add to Cart