RIE156320659 Rieke Metals, Inc.

4-Acetoxy-4'-phenoxybenzophenone

  • MDL number MFCD07782984
  • Cas Number : 890099-60-2
  • Molecular Weight : 332.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5177-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-28
Availability details for
Quantity
  • Availability
  • Add to Cart