RIE156320660 Rieke Metals, Inc.

4-Acetoxy-3'-nitrobenzophenone

  • MDL number MFCD07698960
  • Cas Number : 890099-62-4
  • Molecular Weight : 285.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(3-nitrobenzoyl)phenyl acetate
Inchi Code 1S/C15H11NO5/c1-10(17)21-14-7-5-11(6-8-14)15(18)12-3-2-4-13(9-12)16(19)20/h2-9H,1H3
Inchi Key XLRLBGPZDRRKTM-UHFFFAOYSA-N

Description

Purity 90%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5177-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-30
Availability details for
Quantity
  • Availability
  • Add to Cart