RIE156320664 Rieke Metals, Inc.

4-Acetoxy-4'-t-butylbenzophenone

  • MDL number MFCD07698964
  • Cas Number : 890099-70-4
  • Molecular Weight : 296.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5177-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-34
Availability details for
Quantity
  • Availability
  • Add to Cart