RIE156320665 Rieke Metals, Inc.

4-Acetoxy-4'-ethylbenzophenone

  • MDL number MFCD07698965
  • Cas Number : 178860-08-7
  • Molecular Weight : 268.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5177-35A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-35A
Availability details for
Quantity
  • Availability
  • Add to Cart