RIE156320669 Rieke Metals, Inc.

4-Acetoxy-4'-hexylbenzophenone

  • MDL number MFCD07698969
  • Cas Number : 890099-79-3
  • Molecular Weight : 324.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5177-35E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-35E
Availability details for
Quantity
  • Availability
  • Add to Cart