RIE156320675 Rieke Metals, Inc.

4-Acetoxy-4'-hexyloxybenzophenone

  • MDL number MFCD07698975
  • Cas Number : 890099-91-9
  • Molecular Weight : 340.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5177-36E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-36E
Availability details for
Quantity
  • Availability
  • Add to Cart