RIE156320679 Rieke Metals, Inc.

4-Acetoxy-2',5'-difluorobenzophenone

  • MDL number MFCD07698979
  • Cas Number : 890099-99-7
  • Molecular Weight : 276.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5177-39

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-39
Availability details for
Quantity
  • Availability
  • Add to Cart