RIE156320681 Rieke Metals, Inc.

4-Acetoxy-3',4'-difluorobenzophenone

  • MDL number MFCD07698981
  • Cas Number : 890100-03-5
  • Molecular Weight : 276.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5177-41

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-41
Availability details for
Quantity
  • Availability
  • Add to Cart