RIE156320682 Rieke Metals, Inc.

4-Acetoxy-3',5'-difluorobenzophenone

  • MDL number MFCD07698982
  • Cas Number : 890100-05-7
  • Molecular Weight : 276.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(3,5-difluorobenzoyl)phenyl acetate
Inchi Code 1S/C15H10F2O3/c1-9(18)20-14-4-2-10(3-5-14)15(19)11-6-12(16)8-13(17)7-11/h2-8H,1H3
Inchi Key BSUYOASASRMVJK-UHFFFAOYSA-N

Description

Purity 90%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5177-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-42
Availability details for
Quantity
  • Availability
  • Add to Cart