RIE156320684 Rieke Metals, Inc.

4-Acetoxy-2',4'-dichlorobenzophenone

  • MDL number MFCD07698984
  • Cas Number : 890100-09-1
  • Molecular Weight : 309.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5177-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-44
Availability details for
Quantity
  • Availability
  • Add to Cart