RIE156320686 Rieke Metals, Inc.

4-Acetoxy-2',6'-dichlorobenzophenone

  • MDL number MFCD07698986
  • Cas Number : 890100-11-5
  • Molecular Weight : 309.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5177-46

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-46
Availability details for
Quantity
  • Availability
  • Add to Cart