RIE156320693 Rieke Metals, Inc.

4-Acetoxy-3',4'-dimethoxybenzophenone

  • MDL number MFCD07698993
  • Cas Number : 890100-25-1
  • Molecular Weight : 300.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5177-53

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-53
Availability details for
Quantity
  • Availability
  • Add to Cart