RIE156320695 Rieke Metals, Inc.

4-Acetoxy-2',3'-dimethylbenzophenone

  • MDL number MFCD07698995
  • Cas Number : 890100-29-5
  • Molecular Weight : 268.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5177-55

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-55
Availability details for
Quantity
  • Availability
  • Add to Cart