RIE156320698 Rieke Metals, Inc.

4-Acetoxy-3',4'-dimethylbenzophenone

  • MDL number MFCD07698998
  • Cas Number : 890100-35-3
  • Molecular Weight : 268.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(3,4-dimethylbenzoyl)phenyl acetate
Inchi Code 1S/C17H16O3/c1-11-4-5-15(10-12(11)2)17(19)14-6-8-16(9-7-14)20-13(3)18/h4-10H,1-3H3
Inchi Key DEXWPUINOVTLOS-UHFFFAOYSA-N

Description

Purity 90%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5177-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-59
Availability details for
Quantity
  • Availability
  • Add to Cart