RIE156320700 Rieke Metals, Inc.

4-Acetoxy-3',4',5'-trifluorobenzophenone

  • MDL number MFCD07699000
  • Cas Number : 890100-39-7
  • Molecular Weight : 294.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5177-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5177-61
Availability details for
Quantity
  • Availability
  • Add to Cart