RIE156320705 Rieke Metals, Inc.

4-n-Butyl-3'-iodobenzophenone

  • MDL number MFCD07775714
  • Cas Number : 64358-32-3
  • Molecular Weight : 364.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5180-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5180-21
Availability details for
Quantity
  • Availability
  • Add to Cart