RIE156320707 Rieke Metals, Inc.

4-n-Butyl-3',5'-dichlorobenzophenone

  • MDL number MFCD09801452
  • Molecular Weight : 307.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-butylphenyl)(3,5-dichlorophenyl)methanone
Inchi Code 1S/C17H16Cl2O/c1-2-3-4-12-5-7-13(8-6-12)17(20)14-9-15(18)11-16(19)10-14/h5-11H,2-4H2,1H3
Inchi Key WXXRERYLESREDN-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5180-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5180-48
Availability details for
Quantity
  • Availability
  • Add to Cart