RIE156320710 Rieke Metals, Inc.

4-tert-Butyl-2'-iodobenzophenone

  • MDL number MFCD07775693
  • Molecular Weight : 364.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5181-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5181-20
Availability details for
Quantity
  • Availability
  • Add to Cart