RIE156320711 Rieke Metals, Inc.

4-tert-Butyl-3'-iodobenzophenone

  • MDL number MFCD07775711
  • Molecular Weight : 364.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-tert-butylphenyl)(3-iodophenyl)methanone
Inchi Code 1S/C17H17IO/c1-17(2,3)14-9-7-12(8-10-14)16(19)13-5-4-6-15(18)11-13/h4-11H,1-3H3
Inchi Key YMVKIUVAWUIGLD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5181-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5181-21
Availability details for
Quantity
  • Availability
  • Add to Cart