RIE156320713 Rieke Metals, Inc.

4-tert-Butyl-3',5'-dichlorobenzophenone

  • MDL number MFCD09801455
  • Molecular Weight : 307.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5181-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5181-48
Availability details for
Quantity
  • Availability
  • Add to Cart