RIE156320715 Rieke Metals, Inc.

3-Bromo-2'-chlorobenzophenone

  • MDL number MFCD09801456
  • Cas Number : 135774-38-8
  • Molecular Weight : 295.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5182-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5182-18
Availability details for
Quantity
  • Availability
  • Add to Cart