RIE156320718 Rieke Metals, Inc.

2,3',5'-Trichlorobenzophenone

  • MDL number MFCD09801460
  • Molecular Weight : 285.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5182-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5182-48
Availability details for
Quantity
  • Availability
  • Add to Cart