RIE156320720 Rieke Metals, Inc.

2-Bromo-4'-chlorobenzophenone

  • MDL number MFCD08063806
  • Cas Number : 99585-64-5
  • Molecular Weight : 295.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5184-17

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5184-17
Availability details for
Quantity
  • Availability
  • Add to Cart