RIE156320723 Rieke Metals, Inc.

3',4-Dichloro-4'-fluorobenzophenone

  • MDL number MFCD09801464
  • Molecular Weight : 269.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5185-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5185-16
Availability details for
Quantity
  • Availability
  • Add to Cart