RIE156320728 Rieke Metals, Inc.

3-Chloro-4-fluoro-3'-iodobenzophenone

  • MDL number MFCD09801469
  • Molecular Weight : 360.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chloro-4-fluorophenyl)(3-iodophenyl)methanone
Inchi Code 1S/C13H7ClFIO/c14-11-7-9(4-5-12(11)15)13(17)8-2-1-3-10(16)6-8/h1-7H
Inchi Key FYTQUYIRBYAYKF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5185-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5185-21
Availability details for
Quantity
  • Availability
  • Add to Cart