RIE156320730 Rieke Metals, Inc.

4-tert-Butyl-3'-chloro-4'-fluorobenzophenone

  • MDL number MFCD09801471
  • Molecular Weight : 290.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-tert-butylphenyl)(3-chloro-4-fluorophenyl)methanone
Inchi Code 1S/C17H16ClFO/c1-17(2,3)13-7-4-11(5-8-13)16(20)12-6-9-15(19)14(18)10-12/h4-10H,1-3H3
Inchi Key VNDGIMUCCZSNNK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5185-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5185-34
Availability details for
Quantity
  • Availability
  • Add to Cart