RIE156320732 Rieke Metals, Inc.

3-Chloro-3',4,5'-trifluorobenzophenone

  • MDL number MFCD09801473
  • Molecular Weight : 270.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5185-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5185-42
Availability details for
Quantity
  • Availability
  • Add to Cart