RIE156320734 Rieke Metals, Inc.

4-Fluoro-3,3',5'-trichlorobenzophenone

  • MDL number MFCD09801475
  • Molecular Weight : 303.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5185-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5185-48
Availability details for
Quantity
  • Availability
  • Add to Cart