RIE156320735 Rieke Metals, Inc.

2,3'-Dichloro-5'-fluorobenzophenone

  • MDL number MFCD09801476
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chloro-5-fluorophenyl)(2-chlorophenyl)methanone
Inchi Code 1S/C13H7Cl2FO/c14-9-5-8(6-10(16)7-9)13(17)11-3-1-2-4-12(11)15/h1-7H
Inchi Key HRKAHFYSUKSSRT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5186-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5186-14
Availability details for
Quantity
  • Availability
  • Add to Cart