RIE156320737 Rieke Metals, Inc.

3-Chloro-5-fluoro-2'-iodobenzophenone

  • MDL number MFCD07775700
  • Molecular Weight : 360.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5186-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5186-20
Availability details for
Quantity
  • Availability
  • Add to Cart