RIE156320739 Rieke Metals, Inc.

3-Chloro-5-fluoro-4'-iodobenzophenone

  • MDL number MFCD07775737
  • Molecular Weight : 360.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chloro-5-fluorophenyl)(4-iodophenyl)methanone
Inchi Code 1S/C13H7ClFIO/c14-10-5-9(6-11(15)7-10)13(17)8-1-3-12(16)4-2-8/h1-7H
Inchi Key SPZOWBZSQSGEPD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5186-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5186-22
Availability details for
Quantity
  • Availability
  • Add to Cart