RIE156320743 Rieke Metals, Inc.

3,4'-Dichloro-3'-fluorobenzophenone

  • MDL number MFCD09801481
  • Molecular Weight : 269.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5187-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5187-15
Availability details for
Quantity
  • Availability
  • Add to Cart