RIE156320747 Rieke Metals, Inc.

4-Bromo-4'-chloro-3'-fluorobenzophenone

  • MDL number MFCD09801485
  • Molecular Weight : 313.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5187-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5187-19
Availability details for
Quantity
  • Availability
  • Add to Cart