RIE156320751 Rieke Metals, Inc.

4-tert-Butyl-4'-chloro-3'-fluorobenzophenone

  • MDL number MFCD09801489
  • Molecular Weight : 290.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5187-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5187-34
Availability details for
Quantity
  • Availability
  • Add to Cart