RIE156320754 Rieke Metals, Inc.

3-Fluoro-3',4,4'-trichlorobenzophenone

  • MDL number MFCD09801492
  • Molecular Weight : 303.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5187-47

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5187-47
Availability details for
Quantity
  • Availability
  • Add to Cart