RIE156320756 Rieke Metals, Inc.

2,3'-Dichloro-4'-methylbenzophenone

  • MDL number MFCD09801494
  • Cas Number : 861307-17-7
  • Molecular Weight : 265.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chloro-4-methylphenyl)(2-chlorophenyl)methanone
Inchi Code 1S/C14H10Cl2O/c1-9-6-7-10(8-13(9)16)14(17)11-4-2-3-5-12(11)15/h2-8H,1H3
Inchi Key RWYQEMSOEFOJMP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5188-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5188-14
Availability details for
Quantity
  • Availability
  • Add to Cart