RIE156320765 Rieke Metals, Inc.

4-tert-Butyl-3'-chloro-4'-methylbenzophenone

  • MDL number MFCD09801503
  • Molecular Weight : 286.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5188-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5188-34
Availability details for
Quantity
  • Availability
  • Add to Cart