RIE156320768 Rieke Metals, Inc.

4-Methyl-3,3',4'-trichlorobenzophenone

  • MDL number MFCD09801506
  • Molecular Weight : 299.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5188-47

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5188-47
Availability details for
Quantity
  • Availability
  • Add to Cart