RIE156320773 Rieke Metals, Inc.

2-Bromo-4'-chloro-3'-methylbenzophenone

  • MDL number MFCD09801511
  • Molecular Weight : 309.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5189-17

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5189-17
Availability details for
Quantity
  • Availability
  • Add to Cart