RIE156320777 Rieke Metals, Inc.

4-Chloro-3'-iodo-3-methylbenzophenone

  • MDL number MFCD09801515
  • Molecular Weight : 356.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5189-21

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5189-21
Availability details for
Quantity
  • Availability
  • Add to Cart